missing translation for 'onlineSavingsMsg'
Learn More

6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one 98.0+%, TCI America™

Catalog No. p-7123441
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42701G 1 g
B42705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42701G Supplier TCI America Supplier No. B42701G
Only null left

Chemical Identifiers

CAS 179688-29-0
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.307
MDL Number MFCD02678087
InChI Key PMQWTUWLIGJTQD-UHFFFAOYSA-N
Synonym 6,7-Bis(2-methoxyethoxy)-4(3H)-quinazolinone, 6,7-Bis(2-methoxyethoxy)-3,4-dihydroquinazolin-4-one, 6,7-Bis(2-methoxyethoxy)-4-hydroxyquinazoline, 6,7-Bis(2-methoxyethoxy)-4-quinazolinol
PubChem CID 2809007
IUPAC Name 6,7-bis(2-methoxyethoxy)-1H-quinazolin-4-one
SMILES COCCOC1=C(C=C2C(=C1)C(=O)N=CN2)OCCOC

Specifications

Melting Point 192°C
Quantity 1 g
Formula Weight 294.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one