missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 192°C |
| Quantity | 1 g |
| Formula Weight | 294.31 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one |
Safety and Handling
| TSCA | No |