missing translation for 'onlineSavingsMsg'
Learn More

6,7,8,9-Tetrahydro-5h-carbazole-3-carboxylic acid, ≥97%, Thermo Scientific™

Chemical Identifiers

CAS 36729-27-8
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.252
MDL Number MFCD00458819
InChI Key OWQQDAGRTDUORV-UHFFFAOYSA-N
Synonym 2,3,4,9-tetrahydro-1h-carbazole-6-carboxylic acid, 1,2,3,4-tetrahydrocarbazole-6-carboxylic acid, 1h-carbazole-6-carboxylic acid, 2,3,4,9-tetrahydro, 5,6,7,8,9-pentahydro-4ah-carbazole-3-carboxylic acid, enamine_005086, 2,3,4,9-tetrahydro-1h-carbazole-6-carboxylicacid, 1,2,3,4-tetrahyrocarbazole-6-carboxylic acid
PubChem CID 729801
IUPAC Name 6,7,8,9-tetrahydro-5H-carbazole-3-carboxylic acid
SMILES C1CCC2=C(C1)C3=C(N2)C=CC(=C3)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MO07745CB
SDS View Documents
Thermo Scientific Chemicals
MO07745CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

36729-27-8
215.252
OWQQDAGRTDUORV-UHFFFAOYSA-N
729801
C1CCC2=C(C1)C3=C(N2)C=CC(=C3)C(=O)O
C13H13NO2
MFCD00458819
2,3,4,9-tetrahydro-1h-carbazole-6-carboxylic acid, 1,2,3,4-tetrahydrocarbazole-6-carboxylic acid, 1h-carbazole-6-carboxylic acid, 2,3,4,9-tetrahydro, 5,6,7,8,9-pentahydro-4ah-carbazole-3-carboxylic acid, enamine_005086, 2,3,4,9-tetrahydro-1h-carbazole-6-carboxylicacid, 1,2,3,4-tetrahyrocarbazole-6-carboxylic acid
6,7,8,9-tetrahydro-5H-carbazole-3-carboxylic acid

Specifications

36729-27-8
97%
C13H13NO2
250mg
OWQQDAGRTDUORV-UHFFFAOYSA-N
6,7,8,9-tetrahydro-5H-carbazole-3-carboxylic acid
729801
97%
97.0
Amber Glass Bottle
MFCD00458819
2,3,4,9-tetrahydro-1h-carbazole-6-carboxylic acid, 1,2,3,4-tetrahydrocarbazole-6-carboxylic acid, 1h-carbazole-6-carboxylic acid, 2,3,4,9-tetrahydro, 5,6,7,8,9-pentahydro-4ah-carbazole-3-carboxylic acid, enamine_005086, 2,3,4,9-tetrahydro-1h-carbazole-6-carboxylicacid, 1,2,3,4-tetrahyrocarbazole-6-carboxylic acid
C1CCC2=C(C1)C3=C(N2)C=CC(=C3)C(=O)O
215.252
215.25
6,7,8,9-Tetrahydro-5h-carbazole-3-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'