missing translation for 'onlineSavingsMsg'
Learn More

[6,6]-Phenyl-C61-butyric Acid Methyl Ester 99.5+%, TCI America™

Catalog No. M2088100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2088100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2088100MG Supplier TCI America Supplier No. M2088100MG

Chemical Identifiers

CAS 160848-22-6
Molecular Formula C72H14O2
Molecular Weight (g/mol) 910.902
MDL Number MFCD07784544
InChI Key MCEWYIDBDVPMES-UHFFFAOYSA-N
Synonym Methyl [6,6]-Phenyl-C61-butyrate, [60]PCBM, PCBM
PubChem CID 53384373
SMILES COC(=O)CCCC1(C23C14C5=C6C7=C8C5=C9C1=C5C%10=C%11C%12=C%13C%10=C%10C1=C8C1=C%10C8=C%10C%14=C%15C%16=C%17C(=C%12C%12=C%17C%17=C%18C%16=C%16C%15=C%15C%10=C1C7=C%15C1=C%16C(=C%18C7=C2C2=C%10C(=C5C9=C42)C%11=C%12C%10=C%177)C3=C16)C%14=C%138)C1=CC=CC=C1

Specifications

Color Black-Yellow
Quantity 100 mg
Formula Weight 910.90
Percent Purity ≥99.5% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material [6,6]-Phenyl-C61-butyric Acid Methyl Ester