missing translation for 'onlineSavingsMsg'
Learn More

6,6-Dimethylfulvene 95.0+%, TCI America™

Catalog No. D27861G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D27861G Supplier TCI America Supplier No. D27861G
Only null left

Chemical Identifiers

CAS 2175-91-9
Molecular Formula C8H10
Molecular Weight (g/mol) 106.168
MDL Number MFCD00011655
InChI Key WXACXMWYHXOSIX-UHFFFAOYSA-N
Synonym 5-Isopropylidene-1,3-cyclopentadiene, 5-(Dimethylmethylene)-1,3-cyclopentadiene
PubChem CID 137467
IUPAC Name 5-propan-2-ylidenecyclopenta-1,3-diene
SMILES CC(=C1C=CC=C1)C

Specifications

Melting Point 3°C
Boiling Point 77°C
Quantity 1 g
UN Number 3295
Formula Weight 106.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 6,6-Dimethylfulvene
TSCA No
Recommended Storage Freezer (-20 °C)