missing translation for 'onlineSavingsMsg'
Learn More

6,6'-Dibromo-9,9'-diphenyl-3,3'-bicarbazole 95.0+%, TCI America™

Catalog No. d4980200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4980200MG 200 mg
D49801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4980200MG Supplier TCI America Supplier No. D4980200MG
Only null left

Chemical Identifiers

CAS 354135-75-4
Molecular Formula C36H22Br2N2
Molecular Weight (g/mol) 642.394
InChI Key VTIPHPNQFUCDAW-UHFFFAOYSA-N
PubChem CID 131852839
IUPAC Name 3-bromo-6-(6-bromo-9-phenylcarbazol-3-yl)-9-phenylcarbazole
SMILES C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC5=C(C=C4)N(C6=C5C=C(C=C6)Br)C7=CC=CC=C7)C8=C2C=CC(=C8)Br

Specifications

Quantity 200 mg
Formula Weight 642.39
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 6,6′-Dibromo-9,9′-diphenyl-3,3′-bicarbazole