missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6,6'-Dibromo-2,2'-bipyridyl 95.0+%, TCI America™
$259.11 - $801.54
Chemical Identifiers
| CAS | 49669-22-9 |
|---|---|
| Molecular Formula | C10H6Br2N2 |
| Molecular Weight (g/mol) | 313.98 |
| MDL Number | MFCD00234587 |
| InChI Key | WZVWSOXTTOJQQQ-UHFFFAOYSA-N |
| PubChem CID | 4407547 |
| IUPAC Name | 6,6'-dibromo-2,2'-bipyridine |
| SMILES | BrC1=CC=CC(=N1)C1=NC(Br)=CC=C1 |
Chemical Identifiers
| 49669-22-9 | |
| 313.98 | |
| WZVWSOXTTOJQQQ-UHFFFAOYSA-N | |
| 6,6'-dibromo-2,2'-bipyridine |
| C10H6Br2N2 | |
| MFCD00234587 | |
| 4407547 | |
| BrC1=CC=CC(=N1)C1=NC(Br)=CC=C1 |
Specifications
| 49669-22-9 | |
| White-Yellow | |
| MFCD00234587 | |
| 6,6′C-Dibromo-2,2′C-bipyridine | |
| BrC1=CC=CC(=N1)C1=NC(Br)=CC=C1 | |
| 313.98 | |
| 313.98 | |
| Crystalline Powder |
| 225°C | |
| C10H6Br2N2 | |
| 1 g | |
| WZVWSOXTTOJQQQ-UHFFFAOYSA-N | |
| 6,6'-dibromo-2,2'-bipyridine | |
| 4407547 | |
| ≥95.0% (GC) | |
| 6,6′-Dibromo-2,2′-bipyridyl |
Safety and Handling
TSCA : No