missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
6-[4-(tert-Butoxycarbonyl)piperazin-1-yl]nicotinic acid, ≥97%, Thermo Scientific™
Chemical Identifiers
CAS
201809-22-5
Molecular Formula
C15H21N3O4
Molecular Weight (g/mol)
307.35
MDL Number
MFCD06245543
InChI Key
PWYGTZUOLAGDNK-UHFFFAOYSA-N
Synonym
6-4-tert-butoxycarbonyl piperazin-1-yl nicotinic acid, 6-4-tert-butoxycarbonyl piperazin-1-yl pyridine-3-carboxylic acid, 4-5-carboxy-pyridin-2-yl-piperazine-1-carboxylic acid tert-butyl ester, 6-4-tert-butoxy carbonyl piperazin-1-yl pyridine-3-carboxylic acid, 6-4-2-methylpropan-2-yl oxycarbonyl piperazin-1-yl pyridine-3-carboxylic acid, n4-boc-6-piperazin-1-ylnicotinic acid, 6-4-tert-butoxycarbonyl piperazino nicotinic acid, 6-4-tert-butoxycarbonyl piperazino-nicotinic acid, 1-piperazinecarboxylicacid,4-5-carboxy-2-pyridinyl-,1-1,1-dimethylethyl ester
PubChem CID
2771823
IUPAC Name
6-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]pyridine-3-carboxylic acid
SMILES
CC(C)(C)OC(=O)N1CCN(CC1)C1=NC=C(C=C1)C(O)=O
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
CAS
201809-22-5
Molecular Weight (g/mol)
307.35
InChI Key
PWYGTZUOLAGDNK-UHFFFAOYSA-N
PubChem CID
2771823
SMILES
CC(C)(C)OC(=O)N1CCN(CC1)C1=NC=C(C=C1)C(O)=O
Molecular Formula
C15H21N3O4
MDL Number
MFCD06245543
Synonym
6-4-tert-butoxycarbonyl piperazin-1-yl nicotinic acid, 6-4-tert-butoxycarbonyl piperazin-1-yl pyridine-3-carboxylic acid, 4-5-carboxy-pyridin-2-yl-piperazine-1-carboxylic acid tert-butyl ester, 6-4-tert-butoxy carbonyl piperazin-1-yl pyridine-3-carboxylic acid, 6-4-2-methylpropan-2-yl oxycarbonyl piperazin-1-yl pyridine-3-carboxylic acid, n4-boc-6-piperazin-1-ylnicotinic acid, 6-4-tert-butoxycarbonyl piperazino nicotinic acid, 6-4-tert-butoxycarbonyl piperazino-nicotinic acid, 1-piperazinecarboxylicacid,4-5-carboxy-2-pyridinyl-,1-1,1-dimethylethyl ester
IUPAC Name
6-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]pyridine-3-carboxylic acid
Specifications
CAS
201809-22-5
CAS Max %
100.0
Packaging
Amber Glass Bottle
MDL Number
MFCD06245543
Synonym
6-4-tert-butoxycarbonyl piperazin-1-yl nicotinic acid, 6-4-tert-butoxycarbonyl piperazin-1-yl pyridine-3-carboxylic acid, 4-5-carboxy-pyridin-2-yl-piperazine-1-carboxylic acid tert-butyl ester, 6-4-tert-butoxy carbonyl piperazin-1-yl pyridine-3-carboxylic acid, 6-4-2-methylpropan-2-yl oxycarbonyl piperazin-1-yl pyridine-3-carboxylic acid, n4-boc-6-piperazin-1-ylnicotinic acid, 6-4-tert-butoxycarbonyl piperazino nicotinic acid, 6-4-tert-butoxycarbonyl piperazino-nicotinic acid, 1-piperazinecarboxylicacid,4-5-carboxy-2-pyridinyl-,1-1,1-dimethylethyl ester
SMILES
CC(C)(C)OC(=O)N1CCN(CC1)C1=NC=C(C=C1)C(O)=O
Molecular Weight (g/mol)
307.35
Formula Weight
307.35
Chemical Name or Material
6-[4-(Tert-butoxycarbonyl)piperazin-1-yl]nicotinic acid
CAS Min %
96.5
Assay Percent Range
97%
Molecular Formula
C15H21N3O4
Quantity
250mg
InChI Key
PWYGTZUOLAGDNK-UHFFFAOYSA-N
IUPAC Name
6-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]pyridine-3-carboxylic acid
PubChem CID
2771823
Percent Purity
97%
Safety and Handling
Safety and Handling
GHS H Statement: Causes serious eye irritation. Harmful if swallowed. Causes skin irritation. May cause respiratory irritation.
GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS
missing translation for 'documents'