missing translation for 'onlineSavingsMsg'
Learn More

6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)chroman, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC32839DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC32839DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

1002727-88-9
260.14
NDSHAELUPJMEBM-UHFFFAOYSA-N
43811034
CC1(C)OB(OC1(C)C)C1=CC2=C(OCCC2)C=C1
C15H21BO3
MFCD12028565
2-chroman-6-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chroman, 2-3,4-dihydro-2h-1-benzopyran-6-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, chroman-6-boronic acid, pinacol ester, amtb667, chroman-6-boronic acid pinacol ester, chroman-6-ylboronic acid pinacol ester, 3,4-dihydro-2h-chromene-6-boronic acid, pinacol ester, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-chromene
2-(3,4-dihydro-2H-chromen-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)chroman
96.5
97%
C15H21BO3
MFCD12028565
NDSHAELUPJMEBM-UHFFFAOYSA-N
2-(3,4-dihydro-2H-chromen-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
43811034
97%
1002727-88-9
100.0
Amber Glass Bottle
1g
2-chroman-6-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chroman, 2-3,4-dihydro-2h-1-benzopyran-6-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, chroman-6-boronic acid, pinacol ester, amtb667, chroman-6-boronic acid pinacol ester, chroman-6-ylboronic acid pinacol ester, 3,4-dihydro-2h-chromene-6-boronic acid, pinacol ester, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-chromene
CC1(C)OB(OC1(C)C)C1=CC2=C(OCCC2)C=C1
260.14
260.14
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents