missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ 6.35 mm/10 mm Reducing Union PTFE
Specifications
Specifications
| Packaging Quantity | 3/Pk. |
| Description | For connecting glass dioxin columns and reversible tubes with 10 mm and/or 6.35 mm stem O.D. |
| Material | PTFE |