missing translation for 'onlineSavingsMsg'
Learn More

6-[3-(Dimethylamino)propoxy]pyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog No. p-4380093
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444970050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC444970050 Supplier Thermo Scientific Chemicals Supplier No. 444970050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 918643-56-8 | Molecular Formula: C16H27BN2O3 | Molecular Weight: 306.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 918643-56-8
Molecular Formula C16H27BN2O3
Molecular Weight (g/mol) 306.21
MDL Number MFCD06797989
InChI Key QWJUJXPYTLOKRL-UHFFFAOYSA-N
Synonym n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propan-1-amine, 2-3-n,n-dimethylamino-propoxy pyridine-5-boronic acid, pinacol ester, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yloxy propan-1-amine, dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yloxy-propyl-amine, dimethy 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-22-yloxy propyl amine, dimethyl 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propyl amine, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl oxy-1-propanamine
PubChem CID 17998906
IUPAC Name N,N-dimethyl-3-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]oxypropan-1-amine
SMILES CN(C)CCCOC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 34°C to 37°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 306.21
Percent Purity 97%
Chemical Name or Material 6-[3-(Dimethylamino)propoxy]pyridine-3-boronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only