missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic Acid 97.0+%, TCI America™
$106.25 - $370.95
Chemical Identifiers
| CAS | 55453-87-7 |
|---|---|
| Molecular Formula | C16H12O4 |
| Molecular Weight (g/mol) | 268.268 |
| MDL Number | MFCD00242952 |
| InChI Key | QFGMXJOBTNZHEL-UHFFFAOYSA-N |
| Synonym | 6H-11-Oxodibenzo[b,e]oxepin-2-acetic Acid, Isoxepac |
| PubChem CID | 41448 |
| IUPAC Name | 2-(11-oxo-6H-benzo[c][1]benzoxepin-2-yl)acetic acid |
| SMILES | C1C2=CC=CC=C2C(=O)C3=C(O1)C=CC(=C3)CC(=O)O |
Chemical Identifiers
| 55453-87-7 | |
| 268.268 | |
| QFGMXJOBTNZHEL-UHFFFAOYSA-N | |
| 41448 | |
| C1C2=CC=CC=C2C(=O)C3=C(O1)C=CC(=C3)CC(=O)O |
| C16H12O4 | |
| MFCD00242952 | |
| 6H-11-Oxodibenzo[b,e]oxepin-2-acetic Acid, Isoxepac | |
| 2-(11-oxo-6H-benzo[c][1]benzoxepin-2-yl)acetic acid |
Specifications
| 55453-87-7 | |
| C16H12O4 | |
| 1 g | |
| 6H-11-Oxodibenzo[b,e]oxepin-2-acetic Acid, Isoxepac | |
| C1C2=CC=CC=C2C(=O)C3=C(O1)C=CC(=C3)CC(=O)O | |
| 268.268 | |
| 268.27 | |
| Crystalline Powder |
| 142°C | |
| MFCD00242952 | |
| 2811 | |
| QFGMXJOBTNZHEL-UHFFFAOYSA-N | |
| 2-(11-oxo-6H-benzo[c][1]benzoxepin-2-yl)acetic acid | |
| 41448 | |
| ≥97.0% (HPLC,T) | |
| 6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic Acid |
Safety and Handling
RTECSNumber : HQ4110000
TSCA : No