missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic Acid 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 142°C |
| Quantity | 5 g |
| UN Number | 2811 |
| Formula Weight | 268.27 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic Acid |
Safety and Handling
| RTECSNumber | HQ4110000 |
| TSCA | No |