missing translation for 'onlineSavingsMsg'
Learn More

6-(1-Pyrrolidinyl)pyridine-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog No. AAH5012603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5012603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5012603 Supplier Thermo Scientific Chemicals Supplier No. H5012603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 261715-39-3 | Molecular Formula: C10H12N2O | Molecular Weight: 176.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 261715-39-3
Molecular Formula C10H12N2O
Molecular Weight (g/mol) 176.219
MDL Number MFCD06200881
InChI Key FFFAXGTZBUMKTI-UHFFFAOYSA-N
Synonym 6-1-pyrrolidinyl nicotinaldehyde, 6-pyrrolidin-1-yl nicotinaldehyde, 6-pyrrolidin-1-yl pyridine-3-carbaldehyde, 6-pyrrolidin-1-yl pyridine-3-carboxaldehyde, 6-1-pyrrolidinyl pyridine-3-carboxaldehyde, 6-pyrrolidin-1-ylnicotinaldehyde, 6-pyrrolidinylpyridine-3-carbaldehyde, 6-1-pyrrolidino pyridine-3-carboxaldehyde, acmc-1cg5q, 6-1-pyrrolidino nicotinaldehyde
PubChem CID 2794775
IUPAC Name 6-pyrrolidin-1-ylpyridine-3-carbaldehyde
SMILES C1CCN(C1)C2=NC=C(C=C2)C=O

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material 6-(1-Pyrrolidinyl)pyridine-3-carboxaldehyde
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only