missing translation for 'onlineSavingsMsg'
Learn More

(5RS)-3-(2-Hydroxyphenyl)-5-phenylcyclohex-2-enone, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022960
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022960 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022960 Supplier Merck Emd Millipore Supplier No. BP888

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 37209-23-7
Flash Point Not Applicable
Molecular Formula C18H16O2
Linear Formula C18H16O2
Quantity 25 mg
Synonym 3-(2-Hydroxyphenyl)-5-phenyl-2-cyclohexen-1-one; Alice's ketone
Formula Weight 264.32
Grade BP