missing translation for 'onlineSavingsMsg'
Learn More

(5R,6S)-(-)-4-Boc-5,6-diphenyl-2-morpholinone, 98%

Catalog No. p-7053532
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6076403 1 g
AAH6076406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6076403 Supplier Thermo Scientific Chemicals Supplier No. H6076403
Only null left

CAS: 112741-49-8 | C21H23NO4 | 353.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112741-49-8
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00074955
InChI Key MRUKRSQUUNYOFK-UHFFFAOYNA-N
Synonym tert-butyl 2r,3s---6-oxo-2,3-diphenyl-4-morpholinecarboxylate, 2r,3s-tert-butyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate, 5s,6r-n-boc-5,6-diphenyl-2-morpholinone, tert-butyl 2r,3s-6-oxo-2,3-diphenylmorpholine-4-carboxylate, 2r,3s---n-boc-6-oxo-2,3-diphenylmorpholine, tert-butyl-2r,3s---6-oxo-2,3-diphenyl-4-morpholinecarboxylate, 4-morpholinecarboxylicacid, 6-oxo-2,3-diphenyl-, 1,1-dimethylethyl ester, 2r,3s, pubchem11707
PubChem CID 981230
IUPAC Name tert-butyl (2R,3S)-6-oxo-2,3-diphenylmorpholine-4-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(=O)OC(C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 206°C (decomposition)
Quantity 1 g
Formula Weight 353.41
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (5R,6S)-(-)-4-Boc-5,6-diphenyl-2-morpholinone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only