CELLTREAT Scientific Products 5 Serological Pipet. Convenient shelf carton allows easy access to pipets and neat storage. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. RNase/DNase Free, Non-pyrogenic. Every case is is printed with the lot number for quality traceability. Packaged sterile, 100 individually wrapped in paper/plastic wrapper per dispensor carton, 200/case.