Learn More
Mtc Bio Transfer pipette, 5ml, large bulb, sterile, 20 per bag, 500/cs
Catalog No. NC1859351
Encompass
This item has a minimum qty of 4 per supplier requirements.These unbreakable, one-piece pipettes eliminate contaminated rubber bulbs, broken glass & exposure to infectious materials. They are made of ultra-clear, low density polyethylene (LDPE) & are ideal for microbiology, blood banking, hematology, immunology, & urinalysis. Sterile pipettes are excellent for tissue culture application. Their low-affinity surface reduces sample, cell or protein loss. Whether you make your selection based on length, build draw, pipette capacity or number of drops per ml, one of these ten designs is likely to be suitable for your application. Some have ultra-fine extended tips (P4113, P4117,P4121, P4122, P4133) that allow for extremely precise dispensing & access into the narrowest vessels.
Catalog No. NC1859351 Supplier Mtc Bio Supplier No. P411114
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This item has a minimum qty of 4 per supplier requirements.These unbreakable, one-piece pipettes eliminate contaminated rubber bulbs, broken glass & exposure to infectious materials. They are made of ultra-clear, low density polyethylene (LDPE) & are ideal for microbiology, blood banking, hematology, immunology, & urinalysis. Sterile pipettes are excellent for tissue culture application. Their low-affinity surface reduces sample, cell or protein loss. Whether you make your selection based on length, build draw, pipette capacity or number of drops per ml, one of these ten designs is likely to be suitable for your application. Some have ultra-fine extended tips (P4113, P4117,P4121, P4122, P4133) that allow for extremely precise dispensing & access into the narrowest vessels.