missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium Nitrate, Technical, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystals and/or Chunks |
| Quantity | 2.5 kg |
| Grade | Technical |
| Packaging | Amber Glass Bottle |
| Chemical Name or Material | Chromium Nitrate, |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.