missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Magnesium Disodium Salt, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Fiber Drum |
| Quantity | 25 kg |
| Molecular Weight (g/mol) | 358.52 |
| Grade | Reagent |
| Insoluble Matter | 5e-05 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.