missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5H-Pyrido[4,3-b]indole 98.0+%, TCI America™
$188.27 - $641.24
Chemical Identifiers
| CAS | 244-69-9 |
|---|---|
| Molecular Formula | C11H8N2 |
| Molecular Weight (g/mol) | 168.20 |
| MDL Number | MFCD00965976 |
| InChI Key | RDMFHRSPDKWERA-UHFFFAOYSA-N |
| Synonym | gamma-Carboline |
| PubChem CID | 130802 |
| IUPAC Name | 5H-pyrido[4,3-b]indole |
| SMILES | N1C2=C(C=CC=C2)C2=C1C=CN=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P2362200MG
|
TCI America
P2362200MG |
200 mg |
Each for $188.27
|
|
|||||
|
P23621G
|
TCI America
P23621G |
1 g |
Each for $641.24
|
|
|||||
Chemical Identifiers
| 244-69-9 | |
| 168.20 | |
| RDMFHRSPDKWERA-UHFFFAOYSA-N | |
| 130802 | |
| N1C2=C(C=CC=C2)C2=C1C=CN=C2 |
| C11H8N2 | |
| MFCD00965976 | |
| gamma-Carboline | |
| 5H-pyrido[4,3-b]indole |
Specifications
| 244-69-9 | |
| C11H8N2 | |
| 200 mg | |
| RDMFHRSPDKWERA-UHFFFAOYSA-N | |
| 5H-pyrido[4,3-b]indole | |
| 130802 | |
| ≥98.0% (GC) | |
| 5H-Pyrido[4,3-b]indole |
| 233°C | |
| MFCD00965976 | |
| gamma-Carboline | |
| N1C2=C(C=CC=C2)C2=C1C=CN=C2 | |
| 168.20 | |
| 168.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : No