Learn More
New England Biolab Ltd 5 DNA Adenylation Kit 10 reactions
Catalog No. 504300
Encompass
The 5' DNA adenylation Kit is a simple and efficient enzymatic method for generating 5' -adenylated DNA. The kit is optimized to produce the adenylated DNA with or without 3' -terminator. The 5 DNA adenylation kit routinely generates greater than 95% conversion of pDNA to AppDNA. This highly efficient process eliminates the need for gel isolation of the product and increases overall yield.
Catalog No. 504300 Supplier New England Biolab Ltd Supplier No. E2610S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The 5' DNA adenylation Kit is a simple and efficient enzymatic method for generating 5' -adenylated DNA. The kit is optimized to produce the adenylated DNA with or without 3' -terminator. The 5 DNA adenylation kit routinely generates greater than 95% conversion of pDNA to AppDNA. This highly efficient process eliminates the need for gel isolation of the product and increases overall yield.