Learn More
Eppendorf™ 5804/5810 Series Four-Place Swinging Bucket Tube Adapters
Fit in buckets supplied with 05-400-94 swinging bucket rotor to accept tubes from 1.5 to 100mL
$635.83 - $690.78
Specifications
| For Use With (Equipment) | A-4-44 swing bucket rotor for 5804/5804R/5810/5810R centrifuge |
|---|
| Catalog Number | Mfr. No. | Bore Size (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Bore Size (Metric) | Price | Quantity & Availability | |||||
|
0540098
|
Eppendorf™
022637568 |
1.6 cm |
Pack of 2 Each for $635.83
|
|
|||||
|
05400106
|
Eppendorf™
022637720 |
4.5 cm |
Pack of 2 Each for $690.78
|
|
|||||
|
05400100
|
Eppendorf™
022637606 |
1.75 cm |
Pack of 2 Each for $635.83
|
|
|||||
|
05400103
|
Eppendorf™
022637665 |
3.1 cm |
Pack of 2 Each for $635.83
|
|
|||||
|
0540097
|
Eppendorf™
022637541 |
1.3 cm |
Pack of 2 Each for $635.83
|
|
|||||
|
0540096
|
Eppendorf™
022637525 |
1.1 cm |
Pack of 2 Each for $635.83
|
|
|||||
Description
Adapters are sold in packs of two. Order the proper number of adapters to fully equip your rotor.
Specifications
| A-4-44 swing bucket rotor for 5804/5804R/5810/5810R centrifuge |