missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ 58-400 Polypropylene Caps, Unlined

Catalog No. 02927142
Change view
Click to view available options
Thread Size (Metric):
24 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
02927142 24 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02927142 Supplier DWK Life Sciences Supplier No. W240545

Case of 1100 unlined white polypropylene caps for bottles with screw cap tops size 58-400.

  • For use with Wheaton screw cap bottles/vials
  • Opening (outer diameter, including threads): 58mm
  • Thread finish: GSI 400 = 1 turn

Specifications

Certification Parameters Screw
Coating PP
Material Polypropylene
Quantity Bottle Caps
Thread Size (Metric) 24 mm
Cell Growth Area 1100/Case
Neck Finish 24 mm
Product Type Screw Cap