missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 55 cal Arc Flash Kit and Voltage Gloves (09)

Catalog No. 170011799
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011799 X-Large
170011767 Small
170011772 Medium
170011777 Large
170011782 2X-Large
170011787 3X-Large
170011792 4X-Large
170011798 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170011799 Supplier National Safety Apparel™ Supplier No. KIT4SC55XL09

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kit includes: 55 cal short coat and bib overall, Arc flash hood with PureView™ faceshield and hard hat, safety glasses, hood bag and gear bag

  • 30% lighter than standard 40 cal kits
  • FR hook and loop front closure
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Hook and loop at side of bib for easy access
  • Made in the USA

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Product Type Arc Flash Kit
Gender Unisex
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Arc Flash Rating 55 cal/cm²
Material Double-Layer TECGEN CC™
Seam Type Stitched
Closure Type FR Hook and Loop
Color Navy
Size X-Large
Suspender Adjustable
Ankle Style Open
Show More Show Less