missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ 50 cal Arc Goggle Hood System

Catalog No. 170012008
Change view
Click to view available options
Arc Flash Rating:
53 cal/cm²
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Arc Flash Rating
170012008 53 cal/cm²
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 170012008 Supplier National Safety Apparel™ Supplier No. KITHP45

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kit Includes: Black OPF blend balaclava and high energy PureView™ arc goggle with anti-fog inner lens

  • Lightweight alternative for 50 cal with 360 degree head protection
  • Provides increased flexibility with excellent breathability
  • Goggles feature baffled top and bottom vents for increased airflow and comfortable elastic strap to secure goggles
  • Balaclava features stretch material keeps head and neck comfortable and when worn with goggles
  • Full facial protection that extends below the neckline
  • Nose covering
  • Made in the USA

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Certifications/Compliance CAT 4, NFPA 70E, ASTM F2178, OSHA 1910.269, ANSI/ISEA Z87.1
Closure Type None
Product Type Goggles
Size Universal
Material Westex™ UltraSoft™, OPF Para-Aramid/OPF Blend
Arc Flash Rating 53 cal/cm²
Seam Type Stitched
Color Navy
Suspender None
Show More Show Less