missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 50 mL Nalgene Oak Ridge Tube Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor

Catalog No. 0100377
Catalog No. 0100377 Supplier Thermo Scientific™ Supplier No. 0100377

Accommodate 50mL Oak Ridge tubes in Thermo Scientific™ Fiberlite™ rotors.

Accommodate 50mL Nalgene Oak Ridge tubes in Thermo Scientific™ Fiberlite™ F13-14x50cy or Fiberlite F15-8x50cy rotors.

Specifications

Type Ridge Tube Adapter
Tube Type 50 mL Nalgene Oak Ridge Tube
Capacity 50 mL
For Use With (Equipment) Fiberlite F13-14x50cy, Fiberlite F15-8x50cy rotors
Description 50mL Nalgene Oak Ridge Tube Adapter