Learn More
Beckman Coulter 30 mm Diameter Conical Tube Adapter, Quantity of Two
Catalog No. NC9565018
Encompass
50mL Conical Tube Adapter for GH3.8 Rotors on Allegra X-15R & X-12 Series (set of 2)
Catalog No. NC9565018 Supplier Beckman Coulter Supplier No. 393266
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
50mL Conical Tube Adapter for GH3.8 Rotors on Allegra X-15R & X-12 Series (set of 2)