Learn More
Chemglass Inc 50mL 2-Neck Flask, Reaction, 14/20 Joint Size, 2mm Glass Stopcock, Airfree, Schlenk
Catalog No. AF053001
Encompass
50mL 2-Neck Flask, Reaction, 14/20 Joint Size, 2mm Glass Stpk, Airfree, Schlenk. Round bottom 2-neck flask having standard taper outer joints, both being of the same size. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve. Side neck can be fitted with a CG-3022 septum stopper for sampling or cannula transfer.
Catalog No. AF053001 Supplier Chemglass Inc Supplier No. AF053001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
50mL 2-Neck Flask, Reaction, 14/20 Joint Size, 2mm Glass Stpk, Airfree, Schlenk. Round bottom 2-neck flask having standard taper outer joints, both being of the same size. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve. Side neck can be fitted with a CG-3022 septum stopper for sampling or cannula transfer.