Learn More
Cellpath Stainless-steel Reusable Base Molds
Catalog No. 22222034
Change view
Click to view available options
Dimensions (L x W x H):
7 x 7 x 5 mm
10 x 24 x 5 mm
14 x 24 x 5 mm
24 x 24 x 5 mm
30 x 24 x 9 mm
37 x 24 x 9 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
22222034 14 x 24 x 5 mm
22222032 7 x 7 x 5 mm
22222037 37 x 24 x 9 mm
22222036 30 x 24 x 9 mm
22222035 24 x 24 x 5 mm
22222033 10 x 24 x 5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22222034 Supplier Cellpath Supplier No. GBB141405A
Only null left
Ideal for prostrate and breast cores and can be used with both grid and slatted processing cassettes for both horizontal and vertical sectioning. Cellpath Stainless-steel Reusable Base Moulds are manufactured from high quality stainless steel that put an end to extensive trimming of wax.
  • Made of reusable Stainless steel
  • Stackable to reduce storage area
  • Internal rounded corners enable easy removal of the wax block
  • Features extended edges prevent tipping and provide grip area for ease of handling
  • Available in wide range of sizes
  • GBB-0714-05A suitable for use with needle and punch biopsies
Specifications
Material Stainless Steel
Dimensions (L x W x H) 14 x 24 x 5 mm
Quantity 5/Pk.