Learn More
Celltreat Scientific Products 500mL Wide Mouth Bottle, Round, PP, Non-sterile, 24/CS
Supplier:  Celltreat Scientific Products 229797
Encompass
CELLTREAT Scientific Products 500mL Wide Mouth Bottle. Round shape and easy-pour wide mouth design. Bottle and cap manufactured from virgin polypropylene for excellent chemical resistance with durable, rigid walls. Features liner-less leak proof screw top closure. Bottle and cap come fully assembled. Ideal for specimen and chemical storage, storage of laboratory reagents and biological buffers and for sample shipment and packaging. Non-sterile; autoclavable. Every case is printed with the lot number for quality traceability. Individually packed in bags, 24/case.
Catalog No. 501535083
Only null left