Learn More
Chemglass Life Sciences 500mL Jacketed Reaction Beaker
Supplier: Chemglass Inc CG110304
500mL Jacketed Reaction Beaker. Designed for use in temperature controlled chemical and biological reactions. Bottom interior surface is conical in shape to help eliminate spinout when used with CG-2005 egg shaped spinbars. Hose connections have an O.D. of 10mm at the largest serration.