missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Proshield™ Basic Lab Coats

Catalog No. 19162607
Change view
Click to view available options
Size:
Medium
Large
5X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19162607 Medium
19162603 5X-Large
19162606 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19162607 Supplier DuPont™ Supplier No. PB212SBUMD003000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disposable labcoats are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications.

  • Made of spunbond meltblown spunbond (SMS)
  • Optimized for comfort, softness and breathability
  • For general maintenance, janitorial, and cleaning

Specifications

Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Material SMS
Color Blue
Closure Type Snaps
Cuff Style Open
Disposable Single-use
Sterility Non-sterile
Resistance Light Liquid Splash, Particulates
Style Standard Length
No. of Pockets 2
Collar Type Traditional
Model ProShield 10
For Use With (Application) General Maintenance, Janitorial, Cleaning
Autoclavable Not Autoclavable
Show More Show Less