Learn More
Chemglass Inc Glas-Col Mantle Extension Support for 5000mL Series O Mantle, 11 1/8in ID
Catalog No. CG1001008
Encompass
Glas-Col Mantle Extension Support for 5000mL Series O Mantle, 11 1/8in ID. This lattice mount support holds spherical and hemispherical Glas-Col Series O mantles up to 5-liter capacity. The steel support, with its fabric strap basket, is strong, handy and durable. Attaches easily to stable ring stands or distillation racks. Supports are painted gray. This mantle support is made from 1/2in diameter rod.
Catalog No. CG1001008 Supplier Chemglass Inc Supplier No. CG1001008
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Glas-Col Mantle Extension Support for 5000mL Series O Mantle, 11 1/8in ID. This lattice mount support holds spherical and hemispherical Glas-Col Series O mantles up to 5-liter capacity. The steel support, with its fabric strap basket, is strong, handy and durable. Attaches easily to stable ring stands or distillation racks. Supports are painted gray. This mantle support is made from 1/2in diameter rod.