GA International Thermal-transfer, xylene and solvent-resistant resin ribbons 1.97" x 243' / 50mm x 74m Black 0.5" / 12.7mm
Catalog No. 5000561952
Xylene and solvent-resistant resin ribbons for thermal-transfer printers. When used with the appropriate label, the ribbon will provide a xylene and chemical-resistant printout. Range of sizes available, ensuring compatibility with most common barcode printer brands.
Catalog No.
5000561952
Supplier
GA InternationalSupplier No.
XAR50X74C0.51JC2
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Xylene and solvent-resistant resin ribbons for thermal-transfer printers. When used with the appropriate label, the ribbon will provide a xylene and chemical-resistant printout. Range of sizes available, ensuring compatibility with most common barcode printer brands.