GA International Thermal-transfer, smudge and alcohol-proof resin ribbons 3.5" x 1476' / 89mm x 450m Black 1" / 25.4mm
Catalog No. 5000561912
Smudge and alcohol proof resin ribbon for thermal-transfer printers. Available in either black or white ink formats, the printout obtained when printing with this ribbon will resist spraying and rubbing with alcohols. Range of sizes available, ensuring compatibility with most common barcode printer brands.
Catalog No.
5000561912
Supplier
GA InternationalSupplier No.
RR89X450C11IZ4
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Smudge and alcohol proof resin ribbon for thermal-transfer printers. Available in either black or white ink formats, the printout obtained when printing with this ribbon will resist spraying and rubbing with alcohols. Range of sizes available, ensuring compatibility with most common barcode printer brands.