GA International Color Laboratory Tape 0.94" x 180' / 24mm x 55m Purple From -80°C to +121°C (-112°F to +250°F)
Catalog No. 5000561750
Color laboratory tape. Available in an assortment of eye-catching colors and various widths, perfect for color coding and general lab use. Can be written with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.
Catalog No.
5000561750
Supplier
GA InternationalSupplier No.
PAT24PU
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Color laboratory tape. Available in an assortment of eye-catching colors and various widths, perfect for color coding and general lab use. Can be written with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.