GA International Color Laboratory Tape 0.5" x 180' / 12.7mm x 55m Orange From -80°C to +121°C (-112°F to +250°F)
Catalog No. 5000561747
Color laboratory tape. Available in an assortment of eye-catching colors and various widths, perfect for color coding and general lab use. Can be written with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.
Catalog No.
5000561747
Supplier
GA InternationalSupplier No.
PAT13OR
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Color laboratory tape. Available in an assortment of eye-catching colors and various widths, perfect for color coding and general lab use. Can be written with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.