missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ 5000 Series Disposable Half-Facepiece Respirators
Preassembled respirators are maintenance-free and require no spare parts
Supplier: 3M™ 7000051847
Includes: Organic vapor cartridge and reusable storage bag.
Description
- Ideally suited for intermittent use, plant shutdowns, and applications where respirators wear out or become difficult to clean in short periods of time
- Color coded for easy size identification
- Can be used with NIOSH-approved 3M filters N95, P95, and P100 for oil and non-oil particles
- For convenience and simplicity, some respirator assemblies are packaged with 3M Filters P95 or P100 along with 3M Filter Retainer 501 or Filter Adapter 502
- Some styles feature acid gas and/or organic vapor protection
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Compliance
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
| NIOSH 42 CFR Part 84 | |
| Thermoplastic Elastomer | |
| Medium | |
| Assembly, cleaning, grinding, machine operation, painting, welding | |
| Organic vapor cartridge, reusable storage bag | |
| 4-point | |
| 0.35 kg |
| Yellow | |
| Half-Mask Respirator | |
| Thermoplastic Elastomer | |
| 3M filters N95, P95, and P100 for oil and non-oil particles | |
| 5203 | |
| 0.78 lb. |