missing translation for 'onlineSavingsMsg'
Learn More

3M™ 5000 Series Disposable Half-Facepiece Respirators

Catalog No. 17674B
Change view
Click to view available options
Color:
Black
Yellow
Size:
Small
Medium
Large
For Use With (Application):
Assembly, cleaning, grinding, machine operation, painting, welding
Spray painting, pesticides, agriculture, construction and transportation
Includes:
Organic vapor cartridge, reusable storage bag
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size For Use With (Application) Includes
17674B Black Medium Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
17674C Black Large Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
17676A Yellow Small Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
17676B Yellow Medium Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
17676C Yellow Large Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
19003453A Black Small Spray painting, pesticides, agriculture, construction and transportation Organic vapor cartridge, reusable storage bag
19003453B Black Medium Spray painting, pesticides, agriculture, construction and transportation Organic vapor cartridge, reusable storage bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17674B Supplier 3M™ Supplier No. 7000126026

Includes

Organic vapor cartridge and reusable storage bag.

Preassembled respirators are maintenance-free and require no spare parts

  • Ideally suited for intermittent use, plant shutdowns, and applications where respirators wear out or become difficult to clean in short periods of time
  • Color coded for easy size identification
  • Can be used with NIOSH-approved 3M filters N95, P95, and P100 for oil and non-oil particles
  • For convenience and simplicity, some respirator assemblies are packaged with 3M Filters P95 or P100 along with 3M Filter Retainer 501 or Filter Adapter 502
  • Some styles feature acid gas and/or organic vapor protection

Certifications

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Compliance

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Black
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Medium
Facepiece Material Thermoplastic Elastomer
For Use With (Application) Assembly, cleaning, grinding, machine operation, painting, welding
For Use With (Equipment) 3M filters N95, P95, and P100 for oil and non-oil particles
Includes Organic vapor cartridge, reusable storage bag
Model 5201
Suspension Type 4-point
Show More Show Less