Learn More
Encon™ 500 Series Goggles
Catalog No. 19048483
Encompass
Change view
Click to view available options
Product Type:
Encon™ 500 Series 503Q Safety Goggles
Catalog No. Product Type
19048483 Encon™ 500 Series 503Q Safety Goggles
1 options
Catalog No. 19048483 Supplier Encon™ Supplier No. 05058201
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Aspheric correction allows distortion-free optics
  • Impact-resistant polycarbonate lens provides 99.99% UV absorption up to 380nm
  • Optional Enfog™ antifog coating
  • Lightweight PVC gray plastic frame with universal fit
  • Indirect ventilation system with labyrinth design protects eyes from liquid, dust, and airborne particulates; allows cooling, continuous airflow
  • Forward tilt allows superior field of vision for wearer working at benchtop
  • Black neoprene strap
Order Info
Cat. No. 19-048-483 includes quick-draw strap retainer.
Certifications
CSA Z94.3-2007 standards.
Compliance
Meet ANSI Z87.1-2010-High Impact
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2010, CSA Z94.3-2007
Description Clear, with Enfog coating and strap retainer
Lens Tint Clear
Product Type Encon™ 500 Series 503Q Safety Goggles
Frame Color Gray
Includes Black Neoprene Strap, Indirect Venting, SVQD2 Side Vent
Frame Material Plastic