Learn More
Electron Microscopy Sciences PetriStickers™ Petri Dish Labels 50-Square Grids
Supplier:  Electron Microscopy Sciences 7063050
Encompass
Stickers for labeling Petri dishes. Label Petri dishes quickly, neatly, and reliably. Labels remain firmly intact in temperatures from 4 degree C to 42 degree C. Instantly form an imprint Petri dish for quick reference, then remove if you wish. PetriStickers™ are 3" or 76 mm diameter.
Catalog No. 50930418