Learn More
DuPont™ ProShield™ 50 Lab Coat NB212S WH
Catalog No. 17897235
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17897235 Small
17897234 Medium
17897233 Large
17897236 X-Large
17897230 2X-Large
17897231 3X-Large
17897232 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17897235 Supplier DuPont™ ProShield™ Supplier No. NB212SWHSM003000
Only null left
50 Lab Coat with collar, open wrists, front snap closure, 2 pockets, and serged seams for reliable protection in industrial use.
  • ProShield™ 50 garments protect against non-hazardous particles, light liquid splash, and aerosols in various industrial applications.
  • Made with a microporous film laminated to a spunbonded polypropylene substrate for durability and comfort.
  • Front snap closures allow easy donning and doffing
  • Enhanced frontal protection with a laydown collar.
  • Open wrists for flexible movement.
  • Two front pockets for holding writing instruments, small tools, or parts.
  • Manufactured without natural rubber latex.
Specifications
Size Small
Gender Unisex
Material Polypropylene, Polyethylene Laminate
Color White
Closure Type Snaps
Cuff Style Open
Disposable Single-use
Sterility Non-sterile
Resistance Non-hazardous Liquids
Style Standard Length
No. of Pockets 2
Latex Free No
Autoclavable Not Autoclavable
Show More Show Less