missing translation for 'onlineSavingsMsg'
Learn More

5-Vinylbicyclo[2.2.1]hept-2-ene (stabilized with BHT) 98.0+%, TCI America™

Catalog No. p-7137910
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V006225ML 25 mL
V0062500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V006225ML Supplier TCI America Supplier No. V006225ML
Only null left

Chemical Identifiers

CAS 3048-64-4
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00080691
InChI Key INYHZQLKOKTDAI-UHFFFAOYSA-N
Synonym 5-vinyl-2-norbornene, 2-vinylnorbornene, vinylnorbornene, 5-vinylnorbornene, 2-vinyl-5-norbornene, 2-norbornene, 5-vinyl, 5-vinylnorborn-2-ene, bicyclo 2.2.1 hept-2-ene, 5-ethenyl, vinylnorbornene van, 5-vinylbicyclo 2.2.1 hept-2-ene
PubChem CID 18273
IUPAC Name 5-ethenylbicyclo[2.2.1]hept-2-ene
SMILES C=CC1CC2CC1C=C2

Specifications

Color Colorless
Boiling Point 141°C
Quantity 25 mL
UN Number 1992
Formula Weight 120.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Vinylbicyclo[2.2.1]hept-2-ene (stabilized with BHT)
EINECSNumber (4)-1474
RTECSNumber RC0350000
TSCA Yes