Learn More
5-Vinyl-2-norbornene, 98%, stabilized
CAS: 3048-64-4 | C9H12 | 120.19 g/mol
Supplier: Thermo Scientific Chemicals 164860025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 5-Vinyl-2-norbornene | |
| 3048-64-4 | |
| 100.0 | |
| 141°C | |
| Authentic | |
| Glass Bottle | |
| 1.4800 to 1.482 | |
| 2.5 L | |
| INYHZQLKOKTDAI-UHFFFAOYSA-N | |
| 5-ethenylbicyclo[2.2.1]hept-2-ene | |
| 100 to 200ppm t-butylcatechol | |
| 120.19 | |
| Liquid |
| 98% | |
| 97.5 | |
| -80°C | |
| 27°C | |
| 97.5% min. (GC) | |
| C9H12 | |
| MFCD00080691 | |
| 5-vinyl-2-norbornene, 2-vinylnorbornene, vinylnorbornene, 5-vinylnorbornene, 2-vinyl-5-norbornene, 2-norbornene, 5-vinyl, 5-vinylnorborn-2-ene, bicyclo 2.2.1 hept-2-ene, 5-ethenyl, vinylnorbornene van, 5-vinylbicyclo 2.2.1 hept-2-ene | |
| C=CC1CC2CC1C=C2 | |
| 120.19 | |
| 18273 | |
| 98% |
Chemical Identifiers
| 3048-64-4 | |
| 120.19 | |
| INYHZQLKOKTDAI-UHFFFAOYSA-N | |
| 18273 | |
| C=CC1CC2CC1C=C2 |
| C9H12 | |
| MFCD00080691 | |
| 5-vinyl-2-norbornene, 2-vinylnorbornene, vinylnorbornene, 5-vinylnorbornene, 2-vinyl-5-norbornene, 2-norbornene, 5-vinyl, 5-vinylnorborn-2-ene, bicyclo 2.2.1 hept-2-ene, 5-ethenyl, vinylnorbornene van, 5-vinylbicyclo 2.2.1 hept-2-ene | |
| 5-ethenylbicyclo[2.2.1]hept-2-ene |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 221-259-8
RTECSNumber : RC0350000
TSCA : TSCA
RUO – Research Use Only