missing translation for 'onlineSavingsMsg'
Learn More

5-(Trimethylsilyl)-1,2,3,4,5-pentamethyl-1,3-cyclopentadiene 95.0+%, TCI America™

Catalog No. p-7136900
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16701ML 1 mL
T16705ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16701ML Supplier TCI America Supplier No. T16701ML

Chemical Identifiers

CAS 87778-95-8
Molecular Formula C13H24Si
Molecular Weight (g/mol) 208.42
MDL Number MFCD00151193
InChI Key WNTWQEUDFDAMBF-UHFFFAOYSA-N
PubChem CID 4338309
IUPAC Name trimethyl-(1,2,3,4,5-pentamethylcyclopenta-2,4-dien-1-yl)silane
SMILES CC1=C(C(C(=C1C)C)(C)[Si](C)(C)C)C

Specifications

Boiling Point 101°C
Quantity 1 mL
Formula Weight 208.42
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-(Trimethylsilyl)-1,2,3,4,5-pentamethyl-1,3-cyclopentadiene
TSCA No
Recommended Storage Refrigerator