missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)indole, 98%

Catalog No. AAH28782ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH28782ME 500 mg
AAH28782MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH28782ME Supplier Thermo Scientific Chemicals Supplier No. H28782ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 100846-24-0 | C9H6F3N | 185.149 g/mol

In 3-Naphthylindole construction by rhodium (II)-catalyzed regioselective direct arylation of indoles with 1-diazonaphthalen-2-(1H)-ones treatment of 5-(trifluoromethyl)indole(1k) or 6-(trifluoromethyl)indole (1l) with 2a afforded the desired products 3k (86%)and 3l (86%).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In 3-Naphthylindole construction by rhodium (II)-catalyzed regioselective direct arylation of indoles with 1-diazonaphthalen-2-(1H)-ones treatment of 5-(trifluoromethyl)indole(1k) or 6-(trifluoromethyl)indole (1l) with 2a afforded the desired products 3k (86%)and 3l (86%).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

Chemical Identifiers

CAS 100846-24-0
Molecular Formula C9H6F3N
Molecular Weight (g/mol) 185.149
MDL Number MFCD03095341
InChI Key LCFDJWUYKUPBJM-UHFFFAOYSA-N
Synonym 5-trifluoromethyl indole, 5-trifluoromethyl-1h-indole, 5-trifluoromethylindole, 1h-indole, 5-trifluoromethyl, zlchem 948, pubchem1712, acmc-1box4, 5-trifluoromethyl-indole
PubChem CID 2763075
IUPAC Name 5-(trifluoromethyl)-1H-indole
SMILES C1=CC2=C(C=CN2)C=C1C(F)(F)F

Specifications

Melting Point 68°C to 71°C
Quantity 500 mg
Formula Weight 185.15
Percent Purity 98%
Chemical Name or Material 5-(Trifluoromethyl)indole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only