missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)-2-pyridone 98.0+%, TCI America™

Catalog No. p-7137271
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
T25395G 5 g
T253925G 25 g
2 options

Catalog No. T25395G

Supplier: TCI America T25395G

Only null left

Chemical Identifiers

CAS 33252-63-0
Molecular Formula C6H4F3NO
Molecular Weight (g/mol) 163.10
MDL Number MFCD00042315
InChI Key BYRJSCNPUHYZQE-UHFFFAOYSA-N
Synonym 2-hydroxy-5-trifluoromethylpyridine, 5-trifluoromethyl pyridin-2-ol, 2-hydroxy-5-trifluoromethyl pyridine, 5-trifluoromethyl-2-pyridinol, 5-trifluoromethyl-pyridin-2-ol, 5-trifluoromethyl-2-pyridone, unii-1ccp81h07b, 5-trifluoromethyl pyridin-2 1h-one, 5-trifluoromethyl-1,2-dihydropyridin-2-one, 5-trifloromethyl-2-1h-pyridinone
PubChem CID 147443
IUPAC Name 5-(trifluoromethyl)-1,2-dihydropyridin-2-one
SMILES FC(F)(F)C1=CNC(=O)C=C1

Specifications

Melting Point 149°C
Quantity 5 g
Formula Weight 163.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(Trifluoromethyl)-2-pyridone