missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Trifluoromethyl)-2-pyridone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 149°C |
| Quantity | 5 g |
| Formula Weight | 163.10 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-(Trifluoromethyl)-2-pyridone |
Safety and Handling
| TSCA | No |