missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethoxy)isatin 98.0+%, TCI America™

Catalog No. T27741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27741G 1 g
T27745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27741G Supplier TCI America Supplier No. T27741G

Chemical Identifiers

CAS 169037-23-4
Molecular Formula C9H4F3NO3
Molecular Weight (g/mol) 231.13
MDL Number MFCD00192524
InChI Key XHAJMVPMNOBILF-UHFFFAOYSA-N
Synonym 5-trifluoromethoxy isatin, 5-trifluoromethoxy indoline-2,3-dione, 5-trifluoromethoxy-1h-indole-2,3-dione, 5-trifluoromethoxyisatin, 1h-indole-2,3-dione, 5-trifluoromethoxy, 5-trifluoromethoxy-2,3-indolinedione, buttpark 34\07-90, acmc-209dz9, d0zr2t
PubChem CID 2732752
IUPAC Name 5-(trifluoromethoxy)-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1OC(F)(F)F)C(=O)C(=O)N2

Specifications

Melting Point 171°C
Quantity 1 g
Formula Weight 231.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(Trifluoromethoxy)isatin